2-Klorfenol ≥99.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
C0241-25G
C0241-500G
TCIAC0241-25GEA
230
NOK
TCIAC0241-25G
TCIAC0241-500G
2-Klorfenol ≥99.0% (by GC, titration analysis)
2-Klorfenol
Formel:
C₆H₅ClO Kokepunkt: 175 °C Smeltepunkt: 9 °C Tetthet: 1,27 (20 °C) Flash punkt: 70°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00002159 CAS-nummer: 95-57-8 EINECS: 202-433-2 FN: 2021 ADR: 6.1,III Merck Indeks: 14,02154 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...