1,2,3-Trifluorbensen ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
T2356-1G
T2356-5G
TCIAT2356-1GEA
220
NOK
TCIAT2356-1G
TCIAT2356-5G
1,2,3-Trifluorbensen ≥98.0% (by GC)
1,2,3-Trifluorbensen
Formel:
C₆H₃F₃ MW: 132,09 g/mol Kokepunkt: 95°C Tetthet: 1,28 g/cm³ (20 °C) Flash punkt: –4°C |
MDL nummer:
MFCD00012013 CAS-nummer: 1489-53-8 FN: 1993 ADR: 3,II |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...