1,3,5-Trifluorbensen ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
T1039-25G
T1039-5G
TCIAT1039-25GEA
2375
NOK
TCIAT1039-25G
TCIAT1039-5G
1,3,5-Trifluorbensen ≥98.0% (by GC)
1,3,5-Trifluorbensen
Formel:
C₆H₃F₃ MW: 132,09 g/mol Kokepunkt: 76°C Tetthet: 1,277 g/cm³ (20 °C) Flash punkt: -7°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00000333 CAS-nummer: 372-38-3 EINECS: 206-751-2 FN: 1993 ADR: 3,II |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...