2,3,4,5-Tetrafluorobenzaldehyde ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
T1897-1G
T1897-5G
TCIAT1897-1GEA
1175
NOK
TCIAT1897-1G
TCIAT1897-5G
2,3,4,5-Tetrafluorobenzaldehyde ≥97.0% (by GC)
2,3,4,5-Tetrafluorobenzaldehyde
Formel:
C₇H₂F₄O MW: 178,09 g/mol Kokepunkt: 82°C |
MDL nummer:
MFCD02683041 CAS-nummer: 16583-06-5 FN: 1989 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...