3-Fluorbensonitril ≥98.0% (by GC)

Leverandør: TCI
Danger

F0219-25G F0219-5G
TCIAF0219-25GEA 1250 NOK
TCIAF0219-25G TCIAF0219-5G
3-Fluorbensonitril ≥98.0% (by GC)
3-Fluorbensonitril
Formel: C₇H₄FN
MW: 121,11 g/mol
Kokepunkt: 183°C
Smeltepunkt: –16 °C
Tetthet: 1,1334 g/cm³ (25 °C)
Flash punkt: 68°C
MDL nummer: MFCD00001797
CAS-nummer: 403-54-3
EINECS: 206-963-5
FN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb62fd526ea80d5838f62878f4a0234c852c09a5&vid=4223e1307857d312464f0a681cf6c2d941a4bd56&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR