4-Fluorbensonitril ≥99.0% (by GC)

Leverandør: TCI
Danger

Synonymer: p-Fluorobenzonitrile

F0221-25G F0221-500G F0221-100G
TCIAF0221-25GEA 610 NOK
TCIAF0221-25G TCIAF0221-500G TCIAF0221-100G
4-Fluorbensonitril ≥99.0% (by GC)
4-Fluorbensonitril
Formel: C₇H₄FN
MW: 121,11 g/mol
Kokepunkt: 188°C
Smeltepunkt: 37…40 °C
Tetthet: ~ 1 g/cm³ (20 °C)
Flash punkt: 65°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001812
CAS-nummer: 1194-02-1
EINECS: 214-784-9
FN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4883016d1205d99d67b6717915808382498b0c03&vid=14c45d04bd60229a615afe639d981e2bfc4824d2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR