2-Fluoro-6-iodobenzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
F0796-1G F0796-5G
TCIAF0796-1GEA 365 NOK
TCIAF0796-1G TCIAF0796-5G
2-Fluoro-6-iodobenzoic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-6-iodobenzoic acid
Formel: C₇H₄FIO₂
MW: 266,01 g/mol
Kokepunkt: 312 °C (1013 hPa)
Smeltepunkt: 123…126 °C
MDL nummer: MFCD00042289
CAS-nummer: 111771-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=408ae7b8d94f04be0a16b6182bfe2f480d0d6259&vid=22ecc7b445faf49afaadd5b7ff094f408747ea70&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 124.0 to 128.0 deg-C