2,3,4,5,6-Pentafluoroaniline ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Aminopentafluorobenzene, Pentafluoroaniline
P0922-10G
P0922-25G
P0922-5G
TCIAP0922-10GEA
715
NOK
TCIAP0922-10G
TCIAP0922-25G
TCIAP0922-5G
2,3,4,5,6-Pentafluoroaniline ≥98.0% (by GC)
2,3,4,5,6-Pentafluoroaniline
Formel:
C₆H₂F₅N MW: 183,08 g/mol Kokepunkt: 153°C Smeltepunkt: 36…37 °C Tetthet: 1,744 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007643 CAS-nummer: 771-60-8 EINECS: 212-234-2 |
Spesifikasjon Testresultater
Appearance | White or Colorless to Light yellow to Light orange powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...