1,3,5-Trifluoro-2-nitrobenzene ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
1-Nitro-2,4,6-trifluorobenzene, 2,4,6-Trifluoronitrobenzene
T1681-25G
T1681-5G
TCIAT1681-25GEA
3725
NOK
TCIAT1681-25G
TCIAT1681-5G
1,3,5-Trifluoro-2-nitrobenzene ≥97.0% (by GC)
1,3,5-Trifluoro-2-nitrobenzene
Formel:
C₆H₂F₃NO₂ MW: 177,08 g/mol Kokepunkt: 172°C Smeltepunkt: 3,5 °C Tetthet: 1,514 g/cm³ (20 °C) Flash punkt: 78°C |
MDL nummer:
MFCD00014687 CAS-nummer: 315-14-0 EINECS: 206-248-8 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...