1,2,3-Trifluoro-5-nitrobenzene ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,2,3-Trifluoro-5-nitrobenzene, 3,4,5-Trifluoronitrobenzene

T2357-25G T2357-5G
TCIAT2357-25GEA 1075 NOK
TCIAT2357-25G TCIAT2357-5G
1,2,3-Trifluoro-5-nitrobenzene ≥98.0% (by GC)
1,2,3-Trifluoro-5-nitrobenzene
Formel: C₆H₂F₃NO₂
MW: 177,08 g/mol
Flash punkt: 75°C
MDL nummer: MFCD00456803
CAS-nummer: 66684-58-0
EINECS: 266-447-0
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c18a89516ee2be1444c8484119fdf85b4f0a61d4&vid=c269d092c5a5ab7780581accc71d04245d45ccef&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR