2,5-Difluoroanisole ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,4-Difluoro-2-methoxybenzene

D3805-25G D3805-5G
TCIAD3805-25GEA 1175 NOK
TCIAD3805-25G TCIAD3805-5G
2,5-Difluoroanisole ≥98.0% (by GC)
2,5-Difluoroanisole
Formel: C₇H₆F₂O
MW: 144,12 g/mol
Kokepunkt: 73°C
Flash punkt: 40°C
MDL nummer: MFCD00042561
CAS-nummer: 75626-17-4
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70442aca47ba823176141613d9718be02c93ebbe&vid=30e4561cbd000db17c42294d43810be33b1b4db7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR