2-Fluoro-4-nitrophenol ≥97.0% (by GC)

Leverandør: TCI
Warning

F0381-25G F0381-5G
TCIAF0381-25GEA 1875 NOK
TCIAF0381-25G TCIAF0381-5G
2-Fluoro-4-nitrophenol ≥97.0% (by GC)
2-Fluoro-4-nitrophenol
Formel: C₆H₄FNO₃
MW: 157,1 g/mol
Smeltepunkt: 118…118,5 °C
MDL nummer: MFCD00051970
CAS-nummer: 403-19-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=964976dabfef8a16b3d48aa275940d4e4906dda4&vid=6a7888058268e7c0e11163d26246b3ca16846773&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 118.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR