3-Nitrophenol ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
m-Nitrophenol
N0217-25G
N0217-500G
N0217-100G
TCIAN0217-25GEA
915
NOK
TCIAN0217-25G
TCIAN0217-500G
TCIAN0217-100G
3-Nitrophenol ≥98.0% (by GC)
3-Nitrophenol
Formel:
C₆H₅NO₃ MW: 139,11 g/mol Kokepunkt: 194°C Smeltepunkt: 94…95 °C Tetthet: 1,485 g/cm³ (20 °C) Flash punkt: ≥ 100 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007240 CAS-nummer: 554-84-7 EINECS: 209-073-5 FN: 1663 ADR: 6.1,III Merck Indeks: 14,06618 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 96.0 to 99.0 deg-C |
Solubility in EtOH (95vol%) | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...