Methyl isobutyrate ≥99.0% (by GC)

Leverandør: TCI
Danger

I0106-25ML I0106-500ML
TCIAI0106-25MLEA 220 NOK
TCIAI0106-25ML TCIAI0106-500ML
Methyl isobutyrate ≥99.0% (by GC)
Methyl isobutyrate
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
Kokepunkt: 92°C
Smeltepunkt: –85°C
Flash punkt: 13°C
MDL nummer: MFCD00008914
CAS-nummer: 547-63-7
EINECS: 208-929-5
FN: 1237
ADR: 3,II
Merck Indeks: 14,06088
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e2247f44fecee2a2eb3cbb931ffbb61522af6fb&vid=3e2247f44fecee2a2eb3cbb931ffbb61522af6fb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR