1-Acetyl-4-piperidone ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
N-Acetyl-4-piperidone
A1398-25G
A1398-5G
TCIAA1398-25GEA
1725
NOK
TCIAA1398-25G
TCIAA1398-5G
1-Acetyl-4-piperidone ≥98.0% (by GC)
1-Acetyl-4-piperidone
Formel:
C₇H₁₁NO₂ MW: 141,17 g/mol Flash punkt: 112°C |
MDL nummer:
MFCD00006190 CAS-nummer: 32161-06-1 EINECS: 250-937-6 |
Spesifikasjon Testresultater
Appearance | Colorless to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...