3-Fluoroanthranilic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Warning
Synonymer: 2-Amino-3-fluorobenzoic acid
F0570-1G F0570-5G
TCIAF0570-1GEA 500 NOK
TCIAF0570-1G TCIAF0570-5G
3-Fluoroanthranilic acid ≥98.0% (by titrimetric analysis)
3-Fluoroanthranilic acid
Formel: C₇H₆FNO₂
MW: 155,13 g/mol
MDL nummer: MFCD01569395
CAS-nummer: 825-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=171d503dcbfb1f9f97b5dc6c8b292ba9d03c4171&vid=171d503dcbfb1f9f97b5dc6c8b292ba9d03c4171&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 185.0 to 189.0 deg-C