Methyl indole-4-carboxylate ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
M1773-1G
M1773-5G
TCIAM1773-1GEA
380
NOK
TCIAM1773-1G
TCIAM1773-5G
Methyl indole-4-carboxylate ≥98.0% (by GC)
Methyl indole-4-carboxylate
Formel:
C₁₀H₉NO₂ MW: 175,19 g/mol |
MDL nummer:
MFCD00191222 CAS-nummer: 39830-66-5 |
Spesifikasjon Testresultater
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...