(R)-(+)-1-(2-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: (R)-1-(2-Naphthyl)ethylamine
N0724-1G N0724-5G
TCIAN0724-1GEA 610 NOK
TCIAN0724-1G TCIAN0724-5G
(R)-(+)-1-(2-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)
(R)-(+)-1-(2-Naphthyl)ethylamine
Formel: C₁₂H₁₃N
MW: 171,24 g/mol
Kokepunkt: 166°C
Smeltepunkt: 53°C
MDL nummer: MFCD00085367
CAS-nummer: 3906-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=456e9befccf324c163261a1f21787466bd2e3f00&vid=456e9befccf324c163261a1f21787466bd2e3f00&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +20.0 to +23.0deg (C=1, MeOH)