4-Chloro-4'-hydroxybenzophenone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
(4-Chlorophenyl)(4-hydroxyphenyl)methanone
C1155-25G
C1155-500G
TCIAC1155-25GEA
445
NOK
TCIAC1155-25G
TCIAC1155-500G
4-Chloro-4'-hydroxybenzophenone ≥98.0% (by GC)
4-Chloro-4'-hydroxybenzophenone
Formel:
C₁₃H₉ClO₂ MW: 232,67 g/mol |
MDL nummer:
MFCD00002357 CAS-nummer: 42019-78-3 EINECS: 255-627-4 |
Spesifikasjon Testresultater
Appearance | White to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 178.0 to 181.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...