3-Iodobenzaldehyde ≥96.0% (by GC)

Leverandør: TCI

I0611-25G I0611-5G
TCIAI0611-25GEA 3325 NOK
TCIAI0611-25G TCIAI0611-5G
3-Iodobenzaldehyde ≥96.0% (by GC)
3-Iodobenzaldehyde
Formel: C₇H₅IO
MW: 232,02 g/mol
Kokepunkt: 125°C
MDL nummer: MFCD00039573
CAS-nummer: 696-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6c7cd53caaf4be8ee48e6627e591f2bd9193a0f&vid=a6c7cd53caaf4be8ee48e6627e591f2bd9193a0f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 56.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR