α-Brom-2-klortoluen ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide

C1443-25G C1443-5G
TCIAC1443-25GEA 1250 NOK
TCIAC1443-25G TCIAC1443-5G
α-Brom-2-klortoluen ≥98.0% (by GC)
α-Brom-2-klortoluen
Formel: C₇H₆BrCl
MW: 205,48 g/mol
Kokepunkt: 130°C
Tetthet: 1,5897 g/cm³ (20 °C)
Flash punkt: 109°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000566
CAS-nummer: 611-17-6
EINECS: 210-257-2
FN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79ee313b0e4e3f82f2f9b3b30bf6233f559e6e89&vid=710dc41cc0a620826835d400b0f720f0f87b7a51&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR