4-brombensyl klorid ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
4-Bromo-α-chlorotoluene, α-Chloro-4-bromotoluene, 1-Bromo-4-(chloromethyl)benzene, 1-(Chloromethyl)-4-bromobenzene
B3042-25G
B3042-5G
TCIAB3042-25GEA
1850
NOK
TCIAB3042-25G
TCIAB3042-5G
4-brombensyl klorid ≥98.0% (by GC)
4-brombensyl klorid
Formel:
C₇H₆BrCl MW: 205,48 g/mol Kokepunkt: 236°C Smeltepunkt: 40…41 °C |
MDL nummer:
MFCD00040867 CAS-nummer: 589-17-3 EINECS: 209-638-6 FN: 3261 ADR: 8,III Merck Indeks: 14,01425 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...