2,4-Difluorbensyl bromid ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
1-(Bromomethyl)-2,4-difluorobenzene, α-Bromo-2,4-difluorotoluene
B1710-25G
B1710-5G
TCIAB1710-25GEA
2725
NOK
TCIAB1710-25G
TCIAB1710-5G
2,4-Difluorbensyl bromid ≥98.0% (by GC)
2,4-Difluorbensyl bromid
Formel:
C₇H₅BrF₂ MW: 207,02 g/mol Kokepunkt: 42 °C (1 -rr) Tetthet: 1,61 g/cm³ (20 °C) Flash punkt: 40°C |
MDL nummer:
MFCD00011649 CAS-nummer: 23915-07-3 EINECS: 245-938-3 FN: 2920 ADR: 8,II |
Spesifikasjon Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...