α-Bromo-4-nitrotoluene ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 4-Nitrobenzyl bromide, 1-(Bromomethyl)-4-nitrobenzene, α-Bromo-p-nitrotoluene

N0181-25G N0181-500G N0181-100G
TCIAN0181-25GEA 500 NOK
TCIAN0181-25G TCIAN0181-500G TCIAN0181-100G
α-Bromo-4-nitrotoluene ≥98.0% (by GC, titration analysis)
α-Bromo-4-nitrotoluene
Formel: C₇H₆BrNO₂
MW: 216,03 g/mol
Kokepunkt: 304 °C (1013 hPa)
Smeltepunkt: 94…96 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007373
CAS-nummer: 100-11-8
EINECS: 202-820-6
FN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbe10bdf455cbdedfb3110de47a7e814680785e8&vid=7b5f25820b7c727a9c03832e7b079fc3e5881a4a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 98.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR