5-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Hydroxy-5-methylbenzaldehyde
M1676-25G
M1676-5G
TCIAM1676-25GEA
1350
NOK
TCIAM1676-25G
TCIAM1676-5G
5-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Methylsalicylaldehyde
Formel:
C₈H₈O₂ MW: 136,15 g/mol Kokepunkt: 218°C Smeltepunkt: 54 °C Tetthet: 1,0913 g/cm³ (59 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00274297 CAS-nummer: 613-84-3 EINECS: 210-357-6 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...