4-Metoksyfenylaceton ≥97.0% (by GC)

Leverandør: TCI

Synonymer: 1-(p-Methoxyphenyl)-2-propanone, 1-(4-Methoxyphenyl)-2-propanone

M0675-100ML M0675-25ML
TCIAM0675-100MLEA 730 NOK
TCIAM0675-100ML TCIAM0675-25ML
4-Metoksyfenylaceton ≥97.0% (by GC)
4-Metoksyfenylaceton
Formel: C₁₀H₁₂O₂
MW: 164,2 g/mol
Kokepunkt: 269°C
Smeltepunkt: –15 °C
Tetthet: 1,07 g/cm³ (20 °C)
Flash punkt: 101°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008773
CAS-nummer: 122-84-9
EINECS: 204-578-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a82b12b6d2931b175e637e0ea60cc9153596c055&vid=062056751ea6a1534953172dc87de3403095329e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR