4-Hydroksybensylaceton ≥99.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Raspberry ketone, 1-(4-Hydroxyphenyl)-3-butanone, 4-(4-Hydroxyphenyl)-2-butanone

H0604-25G H0604-250G
TCIAH0604-25GEA 325 NOK
TCIAH0604-25G TCIAH0604-250G
4-Hydroksybensylaceton ≥99.0% (by GC)
4-Hydroksybensylaceton
Formel: C₁₀H₁₂O₂
MW: 164,2 g/mol
Kokepunkt: 292 °C (1013 hPa)
Smeltepunkt: 83…84 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002394
CAS-nummer: 5471-51-2
EINECS: 226-806-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2593bb4437394bf4f038f71e591c24e90006b48&vid=47381a1e1d8721b197505d0b9c52395cc137a7bb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 82.0 to 85.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR