4-Oktylbensoe syre ≥97.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
O0137-1G
O0137-25G
O0137-5G
TCIAO0137-1GEA
460
NOK
TCIAO0137-1G
TCIAO0137-25G
TCIAO0137-5G
4-Oktylbensoe syre ≥97.0% (by GC, titration analysis)
4-Oktylbensoe syre
Formel:
C₁₅H₂₂O₂ MW: 234,34 g/mol Kokepunkt: 362 °C (1013 hPa) Smeltepunkt: 100…102 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00042649 CAS-nummer: 3575-31-3 EINECS: 222-692-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Mesomorphic range | 95.0 to 117.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...