3-Methylanthranil acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 2-Amino-3-methylbenzoic acid

A1569-25G A1569-5G
TCIAA1569-25GEA 1150 NOK
TCIAA1569-25G TCIAA1569-5G
3-Methylanthranil acid ≥98.0% (by titrimetric analysis)
3-Methylanthranil acid
Formel: C₈H₉NO₂
MW: 151,16 g/mol
Kokepunkt: 315 °C (1013 hPa)
Smeltepunkt: 174…175 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007745
CAS-nummer: 4389-45-1
EINECS: 224-505-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=560e97211c4b2b5cfe01f64c34b45697eb944c55&vid=39d01656bec99dcfbdafe5a565a2b6eef05c69db&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR