4-Nitro-m-toluensyre ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Methyl-4-nitrobenzoic acid, 4-Nitro-m-toluic acid
M0565-25G
M0565-500G
TCIAM0565-25GEA
220
NOK
TCIAM0565-25G
TCIAM0565-500G
4-Nitro-m-toluensyre ≥98.0% (by HPLC, titration analysis)
4-Nitro-m-toluensyre
Formel:
C₈H₇NO₄ MW: 181,15 g/mol Kokepunkt: 356 °C (1013 hPa) Smeltepunkt: 212…214 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007168 CAS-nummer: 3113-71-1 EINECS: 221-479-4 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 216.0 to 219.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...