6-Nitro-m-toluensyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 2-Nitro-5-methylbenzoic acid, 5-Methyl-2-nitrobenzoic acid

N0470-25G N0470-5G
TCIAN0470-25GEA 810 NOK
TCIAN0470-25G TCIAN0470-5G
6-Nitro-m-toluensyre ≥98.0% (by titrimetric analysis)
6-Nitro-m-toluensyre
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Kokepunkt: 364 °C (1013 hPa)
Smeltepunkt: 137…138 °C
MDL nummer: MFCD00007368
CAS-nummer: 3113-72-2
EINECS: 221-481-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89064f7a63c40d678652cc7d7a231a16cdf56e2d&vid=e504b7318ff49846fb36421e37fa6d1246068544&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 133.0 to 137.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR