2,3-Diklortoluen ≥98.0% (by GC)

Leverandør: TCI

D1805-25G D1805-500G
TCIAD1805-25GEA 525 NOK
TCIAD1805-25G TCIAD1805-500G
2,3-Diklortoluen ≥98.0% (by GC)
2,3-Diklortoluen
Formel: C₇H₆Cl₂
MW: 161,03 g/mol
Kokepunkt: 208°C
Smeltepunkt: 6 °C
Tetthet: 1,228 g/cm³ (20 °C)
Flash punkt: 83°C
MDL nummer: MFCD00000546
CAS-nummer: 32768-54-0
EINECS: 251-203-8
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55bea58dc1c7fac49611a9734378b560d2811efa&vid=7c8ec16919b2e14e3e2de4abc7b12a7e77112ad9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR