2-klortoluen ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: o-Chlorotoluene

C0295-25G C0295-500G
TCIAC0295-25GEA 350 NOK
TCIAC0295-25G TCIAC0295-500G
2-klortoluen ≥98.0% (by GC)
2-klortoluen
Formel: C₇H₇Cl
MW: 126,59 g/mol
Kokepunkt: 159°C
Smeltepunkt: -35°C
Tetthet: 1,0825 g/cm³ (20 °C)
Flash punkt: 52°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000562
CAS-nummer: 95-49-8
EINECS: 202-424-3
FN: 2238
ADR: 3,III
Merck Indeks: 14,02171
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d279b894a98717db018b29674a399072601f69f5&vid=14606ffc23861996a80d3d229a7e4cbe745163c4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR