α-klor-2-fluortoulen ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-Fluorobenzyl chloride, 1-(Chloromethyl)-2-fluorobenzene, α-Chloro-o-fluorotoulene

F0211-25G F0211-5G
TCIAF0211-25GEA 380 NOK
TCIAF0211-25G TCIAF0211-5G
α-klor-2-fluortoulen ≥98.0% (by GC)
α-klor-2-fluortoulen
Formel: C₇H₆ClF
MW: 144,58 g/mol
Kokepunkt: 175°C
Tetthet: 1,21 g/cm³ (20 °C)
Flash punkt: 57°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00000892
CAS-nummer: 345-35-7
EINECS: 206-460-0
FN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acbed9fd315bee68ca7cc5a2997f20103e4dd2f2&vid=835e17b9d5838c4be85e06350986b9e4103813cf&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR