4-Klor-2-nitrotoluen

Leverandør: TCI
Warning

C0231-25G C0231-500G
TCIAC0231-25GEA 220 NOK
TCIAC0231-25G TCIAC0231-500G
4-Klor-2-nitrotoluen
4-Klor-2-nitrotoluen
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Kokepunkt: 115°C
Smeltepunkt: 38…39 °C
Tetthet: 1,2559 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007215
CAS-nummer: 89-59-8
EINECS: 201-921-2
FN: 3457
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8361fe7cf73cd2380a29a2514ff03cd8c826b9aa&vid=c1632c1518af4de26ff1b086e5bd7d4fa581ba09&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %
Melting point 36.0 to 39.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR