3-Methoxybenzyl chloride ≥96.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
1-(Chloromethyl)-3-methoxybenzene, 3-(Chloromethyl)anisole
M0820-25G
M0820-5G
TCIAM0820-25GEA
785
NOK
TCIAM0820-25G
TCIAM0820-5G
3-Methoxybenzyl chloride ≥96.0% (by GC)
3-Methoxybenzyl chloride
Formel:
C₈H₉ClO MW: 156,61 g/mol Kokepunkt: 125°C Tetthet: 1,07 g/cm³ (20 °C) Flash punkt: 101°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00000907 CAS-nummer: 824-98-6 EINECS: 212-541-1 FN: 3265 ADR: 8,II |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...