Benzotrifluorid ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: α,α,α-Trifluorotoluene, Solvent BTF

T0439-25G T0439-500G
TCIAT0439-25GEA 220 NOK
TCIAT0439-25G TCIAT0439-500G
Benzotrifluorid ≥98.0% (by GC)
Benzotrifluorid
Formel: C₇H₅F₃
MW: 146,11 g/mol
Kokepunkt: 102°C
Smeltepunkt: –29°C
Tetthet: 1,37 g/cm³ (20 °C)
Flash punkt: 12°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000372
CAS-nummer: 98-08-8
EINECS: 202-635-0
FN: 2338
ADR: 3,II
Merck Indeks: 14,01110
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c44f88a920ab837c235d634041f2c3296b60da8&vid=4a256449a75fd1b7cae234dd454b8a4b566912ac&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR