Perfluortoluen ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
Octafluorotoluene, Pentafluoro(trifluoromethyl)benzene, 1,2,3,4,5-Pentafluoro-6-(trifluoromethyl)benzene, α,α,α,2,3,4,5,6-Octafluorotoluene
P0856-25G
P0856-5G
TCIAP0856-25GEA
1100
NOK
TCIAP0856-25G
TCIAP0856-5G
Perfluortoluen ≥98.0% (by GC)
Perfluortoluen
Formel:
C₇F₈ MW: 236,06 g/mol Kokepunkt: 104°C Smeltepunkt: –65,6 °C Tetthet: 1,67 g/cm³ (20 °C) Flash punkt: 20°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00000375 CAS-nummer: 434-64-0 EINECS: 207-104-7 FN: 1993 ADR: 3,II |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...