(4-Fluorfenyl)aceton ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 4-Fluorophenylacetone

F0535-25G F0535-5G
TCIAF0535-25GEA 930 NOK
TCIAF0535-25G TCIAF0535-5G
(4-Fluorfenyl)aceton ≥98.0% (by GC)
(4-Fluorfenyl)aceton
Formel: C₉H₉FO
MW: 152,17 g/mol
Kokepunkt: 130°C
Tetthet: 1,13 g/cm³ (20 °C)
Flash punkt: 91°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000362
CAS-nummer: 459-03-0
EINECS: 207-284-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea1a535d95697b133f9624a2724d4a70a6b6b41e&vid=74cccb2593da9e52f79023b9d231541da0163309&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR