4-Etylanilin ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Danger

E0061-25ML E0061-500ML
TCIAE0061-25MLEA 420 NOK
TCIAE0061-25ML TCIAE0061-500ML
4-Etylanilin ≥99.0% (by GC, titration analysis)
4-Etylanilin
Formel: C₈H₁₁N
MW: 121,18 g/mol
Kokepunkt: 214°C
Smeltepunkt: –5 °C
Tetthet: 0,96875 g/cm³ (20 °C)
Flash punkt: 85°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007921
CAS-nummer: 589-16-2
EINECS: 209-637-0
FN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40605df3352d5a451bd8b38d34548d8572df2ee2&vid=e18f38197add7141583dde4f979bb54c22fd4e48&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR