1-tert-Butyl-4-nitrobenzene ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
1-(tert-Butyl)-4-nitrobenzene
B2149-25G
B2149-5G
TCIAB2149-25GEA
2000
NOK
TCIAB2149-25G
TCIAB2149-5G
1-tert-Butyl-4-nitrobenzene ≥97.0% (by GC)
1-tert-Butyl-4-nitrobenzene
Formel:
C₁₀H₁₃NO₂ MW: 179,22 g/mol |
MDL nummer:
MFCD00041253 CAS-nummer: 3282-56-2 EINECS: 221-922-1 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...