2-Ethylfenol ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
1-Ethyl-2-hydroxybenzene
E0160-25ML
E0160-500ML
E0160-100ML
TCIAE0160-25MLEA
500
NOK
TCIAE0160-25ML
TCIAE0160-500ML
TCIAE0160-100ML
2-Ethylfenol ≥98.0% (by GC)
2-Ethylfenol
Formel:
C₈H₁₀O MW: 122,17 g/mol Kokepunkt: 204,5 °C (1013 hPa) Smeltepunkt: –18 °C Tetthet: 1,0371 g/cm³ (0 °C) Flash punkt: 78°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00002249 CAS-nummer: 90-00-6 EINECS: 201-958-4 FN: 3145 ADR: 8,III Merck Indeks: 14,03839 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...