4'-Hydroxy-3'-methylacetophenone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
H0791-25G
H0791-5G
TCIAH0791-25GEA
1550
NOK
TCIAH0791-25G
TCIAH0791-5G
4'-Hydroxy-3'-methylacetophenone ≥98.0% (by GC)
4'-Hydroxy-3'-methylacetophenone
Formel:
C₉H₁₀O₂ MW: 150,18 g/mol Kokepunkt: 180°C |
MDL nummer:
MFCD00002231 CAS-nummer: 876-02-8 EINECS: 212-880-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 107.0 to 110.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...