4'-Fluoro-3'-nitroacetophenone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
F0777-25G
F0777-5G
TCIAF0777-25GEA
3100
NOK
TCIAF0777-25G
TCIAF0777-5G
4'-Fluoro-3'-nitroacetophenone ≥98.0% (by GC)
4'-Fluoro-3'-nitroacetophenone
Formel:
C₈H₆FNO₃ MW: 183,14 g/mol Kokepunkt: 120°C |
MDL nummer:
MFCD00115369 CAS-nummer: 400-93-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...