2,3-Dimethylbenzoic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Warning
D2221-25G D2221-5G
TCIAD2221-25GEA 1275 NOK
TCIAD2221-25G TCIAD2221-5G
2,3-Dimethylbenzoic acid ≥98.0% (by titrimetric analysis)
2,3-Dimethylbenzoic acid
Formel: C₉H₁₀O₂
MW: 150,18 g/mol
Kokepunkt: 282 °C (1013 hPa)
Smeltepunkt: 143…147 °C
Tetthet: 1,283 g/cm³ (20 °C)
MDL nummer: MFCD00002479
CAS-nummer: 603-79-2
EINECS: 210-058-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4cee24e5229a4358985595e479f3373ea973118&vid=9666d03e6e0345117e9e637de7ef52e67fd9df83&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 144.0 to 147.0 deg-C