α-Klor-p-xylen ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: alpha-Chloro-p-xylene, 4-Methylbenzyl chloride

C0312-25G C0312-500G
TCIAC0312-25GEA 220 NOK
TCIAC0312-25G TCIAC0312-500G
α-Klor-p-xylen ≥98.0% (by GC)
α-Klor-p-xylen
Formel: C₈H₉Cl
MW: 140,61 g/mol
Kokepunkt: 200 °C (1013 hPa)
Smeltepunkt: 5 °C
Tetthet: 1,066 g/cm³ (20 °C)
Flash punkt: 97°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00000919
CAS-nummer: 104-82-5
EINECS: 203-241-1
FN: 3265
ADR: 8,III
Merck Indeks: 14,10091
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed63e7885d222b58abc168c7f9b12b55ed14783a&vid=446dc86687526998c6a216fb02db21fd8d6a716b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR