2,4-Xylenol ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 4-Hydroxy-3-xylene, 2,4-Dimethylphenol, 2,4-Xylenol

D0774-25G D0774-500G
TCIAD0774-25GEA 220 NOK
TCIAD0774-25G TCIAD0774-500G
2,4-Xylenol ≥98.0% (by GC)
2,4-Xylenol
Formel: C₈H₁₀O
MW: 122,17 g/mol
Kokepunkt: 210,9 °C (1013 hPa)
Smeltepunkt: 25 °C
Tetthet: 1,016 g/cm³ (25 °C)
Flash punkt: 80 - 95 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002233
CAS-nummer: 105-67-9
EINECS: 203-321-6
FN: 3430
ADR: 6.1,II
Merck Indeks: 14,10082
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85b2173f97cf3fcfd12b66fff278fe903dd572a5&vid=3c75c9552405e5e3a84839f5a3a713fd0d3e470e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR