Norbornan ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Bicyclo[2.2.1]heptane

N0517-10G N0517-25G
TCIAN0517-10GEA 1950 NOK
TCIAN0517-10G TCIAN0517-25G
Norbornan ≥98.0% (by GC)
Norbornan
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd94162238a9280d5f850e3419377e24cf71a1f2&vid=e1f67bc611471f7bcce995529738d7f9bacdd7f5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 88.0 deg-C
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR