4-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Quinoline-4-carboxylic acid, NSC 13138
Q0065-1G
Q0065-25G
Q0065-5G
TCIAQ0065-1GEA
380
NOK
TCIAQ0065-1G
TCIAQ0065-25G
TCIAQ0065-5G
4-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
4-Quinolinecarboxylic acid
Formel:
C₁₀H₇NO₂ MW: 173,17 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00006782 CAS-nummer: 486-74-8 EINECS: 207-640-1 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...