2-Aminoquinoline ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Quinolin-2-ylamine

A0417-1G A0417-5G
TCIAA0417-1GEA 865 NOK
TCIAA0417-1G TCIAA0417-5G
2-Aminoquinoline ≥97.0% (by GC, titration analysis)
2-Aminoquinoline
Formel: C₉H₈N₂
MW: 144,18 g/mol
Kokepunkt: 305 °C (1013 hPa)
Smeltepunkt: 130 °C
MDL nummer: MFCD00038079
CAS-nummer: 580-22-3
EINECS: 209-458-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cac925c62d91b5bbceb3b15f9f2af5d94a2927df&vid=9c45776c9c919172a56658ff5252e85b8eeef938&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 127.0 to 132.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR